missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9999 LED Mini Light
Click to view available options
Black
Description
- Adjustable LED light ensures optimum vision under poor light conditions
- Easily attaches to the side arms of X-Fit and X-Fit Pro safety spectacles
- Includes matching left and right adapter
Specifications
Color | Black |
Product Type | LED Light |
For Use With (Equipment) | X-Fit and X-Fit Pro Safety Spectacles |
For Use With (Application) | Poor Lighting Conditions |
Includes | Left and Right Adapters |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'