missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9958 Headbands
Click to view available options
For Use With (Equipment):
All Uvex™ Safety Spectacles
Uvex™ 9190 i-3 and i-3s Safety Spectacles
Uvex™ 9199 X-Fit and X-Fit Pro Safety Spectacles
Description
- For use with Uvex™ Safety Spectacles
Specifications
Width (Metric) | 10 mm |
Description | Individual width adjustment |
Color | Black |
Product Type | Headband |
For Use With (Equipment) | Uvex™ 9199 X-Fit and X-Fit Pro Safety Spectacles |
Model | 9958 |
Material | Polyester, Nylon |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'