Learn More
Greiner Bio-One 96-well Plate Frame for C8 Single-Break Strips
GREENER_CHOICE
Product Code. 13005113
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 400
Product Code. 13005113
Brand: Greiner Bio-One 705185
Please to purchase this item. Need a web account? Register with us today!
For use with 96-well microplate. Greiner Bio-One 96-Well Plate Frame for C8 Single-Break Strip holds microplate in place.
  • 1 x 8 Break-apart C-Bottom
  • Flat w/curved edge
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Array 8 x 12
Color White
Material Polystyrene
No. of Wells 96
Sterility Non-sterile