Learn More
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Suitable for tra
Certifications
DIN ISO 9001
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Packaging | Inner bags of 10 |
| Binding Type | Medium Binding |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Microlon 200 |
| Well Shape | Conical Bottom (V-well) |
| Show More |