missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Product Code. p-9045032 Shop All Corning Products
Click to view available options
Packaging:
20/Pk., 100/Cs.
4/Pk
Unit Size:
Pack of 100
Pack of 16
This item is not returnable. View return policy

Product Code. 15368271

Brand: Corning™ 4680

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Half-Area
    Lid With Lid
    Array 8 x 12
    Packaging 4/Pk
    Binding Type TC-Treated
    Color Black, Clear Bottom
    For Use With (Equipment) For high content cell-based assays using imaging systems
    Material Cycloolefin Copolymer
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    No. per Case 16/Cs.
    No. per Pack 4/Pk.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.