missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9175 Skyguard NT Replacement Frame
Click to view available options
Replacement Frame
Description
- Firmly molded TPU overlay construction
- Soft component is flexible with closed pores for an individualized fit
- For use with Skyguard NT safety glasses
Specifications
Certifications/Compliance | EN 166:2001 |
Color | Blue, Gray |
Product Type | Replacement Frame |
For Use With (Equipment) | Syguard NT Safety Glasses |
Disposable | Reusable |
Frame Material | TPU Overlay |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'