Learn More
Tocris Bioscience™ 9-Phenanthrol
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
9-Phenanthrol is a selective TRPM4 blocker (IC50 = 20 μM in HEK293 cells). Exhibits no effect on CFTR or TRPM5 (at 0.25 and 1 mM respectively). Abolishes arrhythmias induced by hypoxia in a mouse heart model.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | 9-Phenanthrol |
| Molecular Weight (g/mol) | 194.23 |
| Inhibitors | 9-Phenanthrol |