Learn More
9-Nitrosancycline Monosulfate, TRC
Specifications
Specifications
| Chemical Name or Material | 9-Nitrosancycline Monosulfate |
| CAS | 2791-13-1 |
| Molecular Formula | C15 H19 N O4 |
| Quantity | 5 mg |
| Synonym | 9-Nitro-6-Demethyl-6-Deoxytetracycline Sulfate, [4S-(4α,4Aα,5Aα,12Aα)]-4-(Dimethylamino)-1,4,4A,5,5A,6,11,12A-Octahydro-3,10,12,12A-Tetrahydroxy-9-Nitro-1,11-Dioxo-2-Naphthacenecarboxamide Sulfate |
| InChI Formula | InChI=1S/C15H19NO4/c1-10(17)16-7-5-11(6-8-16)15(19)13-4-3-12(20-2)9-14(13)18/h3-4,9,11,18H,5-8H2,1-2H3 |
| SMILES | COc1ccc(C(=O)C2CCN(CC2)C(=O)C)c(O)c1 |
| IUPAC Name | 1-[4-(2-hydroxy-4-methoxybenzoyl)piperidin-1-yl]ethanone |
| Molecular Weight (g/mol) | 277.316 |
| Formula Weight | 277.131 |