missing translation for 'onlineSavingsMsg'
Learn More

9-Methylcarbazole-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15419465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419465 1 g 1g
15429465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419465 Supplier Thermo Scientific Alfa Aesar Supplier No. H51952.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217891-71-8
Molecular Formula C19H22BNO2
Molecular Weight (g/mol) 307.20
MDL Number MFCD16294552
InChI Key AVCKYKVGQYBADD-UHFFFAOYSA-N
Synonym 9-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-9h-carbazole, 9-methylcarbazole-3-boronic acid pinacol ester, 9-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl carbazole, amtb254, 9-methyl-9h-carbazole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9-methyl-9h-carbazole
PubChem CID 56924967
SMILES CN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Beilstein 21268368
IUPAC Name 9-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Formula Weight 307.2
Percent Purity 95%
Chemical Name or Material 9-Methylcarbazole-3-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.