Learn More
9,9-Bis (4-Hydroxyphenyl) Fluorene, TRC
Specifications
Specifications
| Chemical Name or Material | Bisphenol FL |
| CAS | 3236-71-3 |
| Molecular Formula | C25 H18 O2 |
| Quantity | 25 g |
| Synonym | Phenol, 4,4'-fluoren-9-ylidenedi- (7CI,8CI), 4,4'-(9H-Fluoren-9-ylidene)bis[phenol], 4,4'-(9H-Fluorene-9,9-diyl)bisphenol, 4,4'-Fluoren-9-ylidenebisphenol, 4,4'-Fluoren-9-ylidenediphenol, 9,9-Bis(4-hydroxyphenyl)-9H-fluorene, 9,9-Bis(4-hydroxyphenyl)fluorene, 9,9-Bis(p-hydroxyphenyl)fluorene, 9,9'-Bis(4-hydroxyphenyl)fluorene, BP-FL 20X, BPF, BPFL, BisP-FL |
| InChI Formula | InChI=1S/C25H18O2/c26-19-13-9-17(10-14-19)25(18-11-15-20(27)16-12-18)23-7-3-1-5-21(23)22-6-2-4-8-24(22)25/h1-16,26-27H |
| SMILES | Oc1ccc(cc1)C2(c3ccc(O)cc3)c4ccccc4c5ccccc25 |
| IUPAC Name | 4-[9-(4-hydroxyphenyl)fluoren-9-yl]phenol |
| Molecular Weight (g/mol) | 350.41 |
| Formula Weight | 350.1307 |