Learn More
9-(2,2-Dicyanovinyl)julolidine, TRC
Specifications
Specifications
| Chemical Name or Material | 9-(2,2-Dicyanovinyl)julolidine |
| CAS | 58293-56-4 |
| Molecular Formula | C16 H15 N3 |
| Quantity | 10 mg |
| Synonym | 2-[(2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-9-yl)methylene]propanedinitrile(Dicyanovinyl)julolidine, 9-(Dicyanovinyl)julolidine, DCVJ, Julolidinemalononitrile, NSC 160064, [(2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-9-yl)methylene]-propanedinitrile |
| InChI Formula | InChI=1S/C16H15N3/c17-10-13(11-18)7-12-8-14-3-1-5-19-6-2-4-15(9-12)16(14)19/h7-9H,1-6H2 |
| SMILES | N#CC(=Cc1cc2CCCN3CCCc(c1)c23)C#N |
| IUPAC Name | 2-(1-azatricyclo[7.3.1.05,13]trideca-5(13),6,8-trien-7-ylmethylidene)propanedinitrile |
| Molecular Weight (g/mol) | 249.31 |
| Formula Weight | 249.127 |