Learn More
9,10-Dehydro Folitixorin Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 9,10-Dehydro Folitixorin Chloride |
| CAS | 804563-04-0 |
| Molecular Formula | C20H22ClN7O6 |
| Quantity | 100 mg |
| Synonym | 3-Amino-8-[4-[[[(1S)-1,3-dicarboxypropyl]amino]carbonyl]phenyl]-2,5,6,6a,7,8-hexahydro-1-oxo-1H-imidazo[1,5-f]pteridin-10-ium Chloride |
| InChI Formula | InChI=1S/C20H21N7O6.ClH/c21-20-24-16-15(18(31)25-20)27-9-26(8-12(27)7-22-16)11-3-1-10(2-4-11)17(30)23-13(19(32)33)5-6-14(28)29;/h1-4,9,12-13H,5-8H2,(H6-,21,22,23,24,25,28,29,30,31,32,33);1H |
| SMILES | O=C1C2=C(NCC3[N+]2=CN(C4=CC=C(C(N[C@H](C(O)=O)CCC(O)=O)=O)C=C4)C3)N=C(N)N1.[Cl-] |
| IUPAC Name | 3-amino-8-(4-(((S)-1,3-dicarboxypropyl)carbamoyl)phenyl)-1-oxo-2,5,6,6a,7,8-hexahydro-1H-imidazo[1,5-f]pteridin-10-ium |
| Molecular Weight (g/mol) | 491.88 |
| Formula Weight | 491.13 |