missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 8-Strip A (Cover for Magnetic Rod) for MPure-32

Product Code. 17712528 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500/Pk.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17712528 500/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17712528 Supplier MP Biomedicals™ Supplier No. 117032148
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

8-Strip A to use with MPure-32 aNAP system. Strips avoid direct contact of magnetic roads with the sample during the mixing step and also the transfer of the magnetic beads. Disposable plastic.

  • To use with MPure-32 aNAP system.
  • Strips avoid:
    • Direct contact of magnetic roads with the sample during the mixing step
    • The transfer of the magnetic beads
  • Disposable plastic
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Tissue, Cell Culture, Bacteria
Format Strips
For Use With (Equipment) MPure-32
Quantity 500/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.