missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxyquinoline, 96%

Product Code. 15490728
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490728 1 g 1g
15410738 250 mg 250mg
15400738 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490728 Supplier Thermo Scientific Alfa Aesar Supplier No. H54430.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 938-33-0 | C10H9NO | 159.19 g/mol

8-Methoxyquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Methoxyquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-33-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00957068
InChI Key ZLKGGEBOALGXJZ-UHFFFAOYSA-N
Synonym quinoline, 8-methoxy, methyl 8-quinolyl ether, ccris 7821, unii-4y56u1unuj, 4y56u1unuj, 8-methoxyquinoline, 8-methoxy quinoline, 8-methoxy-quinoline, acmc-209rns, ksc268q2n
PubChem CID 70310
IUPAC Name 8-methoxyquinoline
SMILES COC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 38°C to 41°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 159.19
Percent Purity 96%
Chemical Name or Material 8-Methoxyquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.