Learn More
8-Hydroxy-3,4-dihydroquinolin-2-oxo-indacaterol, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Hydroxy-3,4-dihydroquinolin-2-oxo-indacaterol |
| CAS | 1403389-05-8 |
| Molecular Formula | C24 H30 N2 O3 |
| Quantity | 5 mg |
| Synonym | (R)-5-(2-((5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino)-1-hydroxyethyl)-8-hydroxy-3,4-dihydroquinolin-2(1H)-one, Indacaterol Impurity 1 |
| InChI Formula | InChI=1S/C24H30N2O3/c1-3-14-9-16-11-18(12-17(16)10-15(14)4-2)25-13-22(28)19-5-7-21(27)24-20(19)6-8-23(29)26-24/h5,7,9-10,18,22,25,27-28H,3-4,6,8,11-13H2,1-2H3,(H,26,29)/t22-/m0/s1 |
| SMILES | CCc1cc2CC(Cc2cc1CC)NC[C@H](O)c3ccc(O)c4NC(=O)CCc34 |
| IUPAC Name | 5-[(1R)-2-[(5,6-diethylindan-2-yl)amino]-1-hydroxy-ethyl]-8-hydroxy-3,4-dihydro-1H-quinolin-2-one |
| Molecular Weight (g/mol) | 394.51 |
| Formula Weight | 394.2256 |