missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. p-7032933
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415215 250 mg 250mg
15405215 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415215 Supplier Thermo Scientific Alfa Aesar Supplier No. H35662.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353258-35-2
Molecular Formula C9H6ClF3N2O3
Molecular Weight (g/mol) 282.603
MDL Number MFCD22682820
InChI Key HBDANCBTRVDEBT-UHFFFAOYSA-N
Synonym 8-chloro-6-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid hydrate
PubChem CID 122361169
IUPAC Name 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid;hydrate
SMILES C1=C(C2=NC(=CN2C=C1C(F)(F)F)C(=O)O)Cl.O

Specifications

Melting Point ∽188°C (decomposition)
Quantity 250 mg
Formula Weight 264.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.