Learn More
8-Bromoguanosine, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Bromoguanosine |
| CAS | 4016-63-1 |
| Molecular Formula | C10 H12 Br N5 O5 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C10H12BrN5O5/c11-9-13-3-6(14-10(12)15-7(3)20)16(9)8-5(19)4(18)2(1-17)21-8/h2,4-5,8,17-19H,1H2,(H3,12,14,15,20)/t2-,4-,5-,8-/m1/s1 |
| SMILES | Nc1nc(O)c2nc(Br)n([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c2n1 |
| IUPAC Name | (2R,3R,4S,5R)-2-(2-amino-8-bromo-6-hydroxypurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| Molecular Weight (g/mol) | 362.14 |
| Formula Weight | 361.0022 |