Learn More
8-Aminoguanine, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Aminoguanine |
| CAS | 28128-41-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5 H6 N6 O |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C5H6N6O/c6-4-8-1-2(9-4)10-5(7)11-3(1)12/h(H6,6,7,8,9,10,11,12) |
| SMILES | NC1=Nc2[nH]c(N)nc2C(=O)N1 |
| IUPAC Name | 2,8-diamino-1,9-dihydropurin-6-one |
| Molecular Weight (g/mol) | 166.14 |
| Formula Weight | 166.0603 |
| Show More |