Learn More
8-(4-Anilino) Bodipy, TRC
Specifications
Specifications
| Chemical Name or Material | 8-(4-Anilino) Bodipy |
| CAS | 321895-93-6 |
| Molecular Formula | C19H20BF2N3 |
| Quantity | 10 mg |
| Synonym | (T-4)-[4-[(3,5-Dimethyl-1H-pyrrol-2-yl-kN)(3,5-dimethyl-2H-pyrrol-2-ylidene-kN)methyl]benzenaminato]difluoroboron |
| InChI Formula | InChI=1S/C19H20BF2N3/c1-11-9-13(3)24-18(11)17(15-5-7-16(23)8-6-15)19-12(2)10-14(4)25(19)20(24,21)22/h5-10H,23H2,1-4H3 |
| SMILES | CC1=CC(=[N+]2C1=C(c3ccc(N)cc3)c4c(C)cc(C)n4[B-]2(F)F)C |
| IUPAC Name | 4-(2,2-difluoro-4,6,10,12-tetramethyl-3-aza-1-azonia-2-boranuidatricyclo[7.3.0.03,7]dodeca-1(12),4,6,8,10-pentaen-8-yl)aniline |
| Molecular Weight (g/mol) | 339.19 |
| Formula Weight | 339.1718 |