Learn More
MP Biomedicals™ 7X™-O-Matic Solution
Product Code. 11459585 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
4 x 1 gal.
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) unitSize
11459585 5 gal. Each
11449585 4 x 1 gal. Pack of 4
11439585 1 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11459585 Supplier MP Biomedicals™ Supplier No. 097667495
Please to purchase this item. Need a web account? Register with us today!
7X™-O-Matic Solution was developed for safe and effective cleaning of laboratory instruments and glassware under pressure, heat and agitation (machine wash)
  • Developed for safe and effective cleaning of laboratory instruments and glassware
  • Formulated to be used in a machine washer to withstand heat, pressure and agitation
  • Powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available, and it is stable for long-term storage
  • Effective and safe
  • Eliminates residue leaving no interfering fluorescence
  • No etch to glass or plastic labware at any concentration
  • Completely drains from glassware in seconds
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
pH 9 to 11
Appearance Clear to Light Yellow
Concentration 7X
Content And Storage 15°C to 30°C
For Use With (Application) Use on Glass, Plastic, benchtop equipment and tubing
Volume (English) 5 gal.
compliance-icons
Product Identifier
  • 7X-O-Matic Cleaning Solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-butoxyethoxy)ethanol, Dioctyl Sulfosuccinate Sodium Salt