missing translation for 'onlineSavingsMsg'
Learn More

7rh, MedChemExpress

Product Code. 16404116
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16404116

Brand: Medchem Express HYU00444

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 7rh (DDR1-IN-2) is a potent inhibitor of discoidin domain receptor 1 (DDR1), with an IC50 of 13.1 nM, and also less potently inhibits DDR2, with an IC50 of 203 nM.

7rh (DDR1-IN-2) is a potent inhibitor of discoidin domain receptor 1 (DDR1), with an IC50 of 13.1 nM, and also less potently inhibits DDR2, with an IC50 of 203 nM.

Specifications

Chemical Name or Material 7rh
CAS 1429617-90-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H29F3N6O
Quantity 25 mg
Synonym DDR1-IN-2
Solubility Information DMSO : 62.5 mg/mL (114.35 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(CN2CCN(C)CC2)=C1)C3=CC=C(CC)C(C#CC4=CN5C(N=C4)=CC=N5)=C3
Molecular Weight (g/mol) 546.59
Formula Weight 546.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.