Learn More
7Beta-Hydroxy-4-cholesten-3-one, TRC
Product Code. 30138198
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138198 10 mg 10mg
30003260 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138198 Supplier Toronto Research Chemicals Supplier No. TRCH82512510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25876-54-4
Molecular Formula C27 H44 O2
Molecular Weight (g/mol) 400.64
Synonym Cholest-4-en-3-one, 7-hydroxy-, (7β)-, Cholest-4-en-3-one, 7β-hydroxy- (8CI), (7β)-7-Hydroxycholest-4-en-3-one, δ4-7β-Hydroxycholestenone, 7β-Hydroxy-4-cholesten-3-one, 7β-Hydroxycholest-4-en-3-one
IUPAC Name (7S,8S,9S,10R,13R,14S,17R)-7-hydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C27H44O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h15,17-18,21-25,29H,6-14,16H2,1-5H3/t18-,21-,22+,23+,24+,25+,26+,27-/m1/s1
Formula Weight 400.3341
Chemical Name or Material 7beta-Hydroxy-4-cholesten-3-one