missing translation for 'onlineSavingsMsg'
Learn More

7ACC1, MedChemExpress

Product Code. 16411476
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
This item is not returnable. View return policy

Product Code. 16411476

Brand: Medchem Express HYD0067

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 7ACC1(DEAC; Coumarin D 1421; D 1421) selectively interfere with lactate fluxes in the lactate-rich tumor microenvironment; inhibits lactate influx but not efflux in tumor cells expressing MCT1 and MCT4 transporters.

7ACC1(DEAC

Specifications

Chemical Name or Material 7ACC1
CAS 50995-74-9
Color Orange
Purity Grade Notes Research
Molecular Formula C14H15NO4
Quantity 200 mg
Synonym DEAC Coumarin D 1421 D 1421
Solubility Information DMSO : 33.33 mg/mL (127.57 mM; Need ultrasonic)
SMILES O=C(C1=CC2=CC=C(N(CC)CC)C=C2OC1=O)O
Molecular Weight (g/mol) 261.27
Formula Weight 261.27
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.