Learn More
TdB Labs AB 70 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies
Product Code. p-200113303
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. 30216397
Brand: TdB Labs AB QD70_10g
Please to purchase this item. Need a web account? Register with us today!
Q-dextran is a positively charged dextran modified with quaternary ammonium groups. It is used in ion exchange chromatography, biochemical assays, and for studying electrostatic interactions with negatively charged biomolecules.
  • Storage and Stability:
    • Store dry at room temperature, protected from moisture and light.
    • Stable for extended periods under recommended storage conditions without loss of charge properties.
  • Specifications
    Quantity 10 g
    Product Type Modified Dextran