Learn More
7-tert-Butyl-6-chloro-3-(2,5-difluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine, TRC
Specifications
Specifications
| Chemical Name or Material | 7-tert-Butyl-6-chloro-3-(2,5-difluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine |
| CAS | 286456-54-0 |
| Molecular Formula | C15 H13 Cl F2 N4 |
| Quantity | 10 mg |
| Synonym | 1,2,4-Triazolo[4,3-b]pyridazine, 6-chloro-3-(2,5-difluorophenyl)-7-(1,1-dimethylethyl)-, 6-Chloro-3-(2,5-difluorophenyl)-7-(1,1-dimethylethyl)-1,2,4-triazolo[4,3-b]pyridazine, 7-tert-Butyl-6-chloro-3-(2,5-difluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine |
| InChI Formula | InChI=1S/C15H13ClF2N4/c1-15(2,3)10-7-12-19-20-14(22(12)21-13(10)16)9-6-8(17)4-5-11(9)18/h4-7H,1-3H3 |
| SMILES | CC(C)(C)c1cc2nnc(c3cc(F)ccc3F)n2nc1Cl |
| IUPAC Name | 7-tert-butyl-6-chloro-3-(2,5-difluorophenyl)-[1,2,4]triazolo[4,3-b]pyridazine |
| Molecular Weight (g/mol) | 322.74 |
| Formula Weight | 322.0797 |