Learn More
7-Phenyl-1-heptanol, 97%
Product Code. 11323226
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323226 5 g 5g
11313226 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11323226 Supplier Thermo Scientific Alfa Aesar Supplier No. L12533.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 3208-25-1 | C13H20O | 192.302 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3208-25-1
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00037315
InChI Key UXMUSYTXSNVRMW-UHFFFAOYSA-N
Synonym benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
PubChem CID 154271
IUPAC Name 7-phenylheptan-1-ol
SMILES C1=CC=C(C=C1)CCCCCCCO
Specifications
Density 0.962
Boiling Point 115°C to 117°C (0.5 mmHg)
Refractive Index 1.508
Quantity 5 g
Beilstein 1865431
Formula Weight 192.3
Percent Purity 97%
Chemical Name or Material 7-Phenyl-1-heptanol