Learn More
7-Methyloctanol, TRC
Specifications
Specifications
| CAS | 2430-22-0 |
| Molecular Formula | C9 H20 O |
| Quantity | 250 mg |
| Synonym | 1-Octanol, 7-methyl- (7CI, 8CI, 9CI, ACI), 7-Methyl-1-octanol (ACI), 7-Methyloctanol, 7-Methyl-1-octyl alcohol |
| InChI Formula | InChI=1S/C9H20O/c1-9(2)7-5-3-4-6-8-10/h9-10H,3-8H2,1-2H3 |
| SMILES | CC(C)CCCCCCO |
| IUPAC Name | 7-methyloctan-1-ol |
| Molecular Weight (g/mol) | 144.255 |
| Formula Weight | 144.151 |
| Chemical Name or Material | 7-Methyloctanol |