Learn More
7-Methyl-1-octyl Alcohol, CDN
Product Code. 30326769
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326769 Supplier CDN Supplier No. CDNX74641G
Please to purchase this item. Need a web account? Register with us today!
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-Methyloctanol
CAS 2430-22-0
Molecular Formula C9 H20 O
Quantity 1 g
Synonym 1-Octanol, 7-methyl- (7 CI, 8 CI, 9 CI, ACI), 7-Methyl-1-octanol (ACI), 7-Methyloctanol, 7-Methyl-1-octyl alcohol
InChI Formula InChI=1 S/C9H20O/c1-9(2)7-5-3-4-6-8-10/h9-10 H,3-8H2,1-2H3
SMILES CC(C)CCCCCCO
IUPAC Name 7-methyloctan-1-ol
Molecular Weight (g/mol) 144.255
Formula Weight 144.151 g/mol
Percent Purity min 98% Chemical Purity