missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide, Thermo Scientific™

Product Code. 15480167
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 15480167

Brand: Thermo Scientific Alfa Aesar J61065.MA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective agonist for D3 dopamine

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74938-11-7
Molecular Formula C16H26BrNO
Molecular Weight (g/mol) 328.294
MDL Number MFCD00153809
InChI Key ODNDMTWHRYECKX-UHFFFAOYSA-N
Synonym 7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-oh-dpat hydrobromide, r +-7-hydroxy-dpat hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, y-7-hydroxy-dpat hydrobromide
PubChem CID 11957566
IUPAC Name 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
SMILES CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br

Specifications

Melting Point 168°C to 170°C
Color White
Quantity 10 mg
Formula Weight 329.29
Physical Form Powder
Chemical Name or Material (+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.