missing translation for 'onlineSavingsMsg'
Learn More

7-Benzyloxy-6-methoxy-4(3H)-quinazolinone, 96%, Thermo Scientific Chemicals

Catalog Number 15418795
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
179688-01-8
C16H14N2O3
282.299
MFCD04115119
ZCUFFSHMOAEEIL-UHFFFAOYSA-N
7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy
10540849
6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one
COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3
This item is not returnable. View return policy
179688-01-8
C16H14N2O3
282.299
MFCD04115119
ZCUFFSHMOAEEIL-UHFFFAOYSA-N
7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy
10540849
6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one
COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 284.31
Percent Purity 96%
Chemical Name or Material 7-Benzyloxy-6-methoxy-4(3H)-quinazolinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'