missing translation for 'onlineSavingsMsg'
Learn More

7-Aminoindole, 97%

Product Code. p-7047156
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11364765

Brand: Thermo Scientific Alfa Aesar L17794.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5192-04-1 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5192-04-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD02093960
InChI Key WTFWZOSMUGZKNZ-UHFFFAOYSA-N
Synonym 7-aminoindole, 7-amino-1h-indole, 1h-indol-7-ylamine, 7-indolamine, indole-7-ylamine, 7-indolylamine, 7-aminoindol, acmc-1ay6v, indol-7-yl amine, 7-aminoindole hplc
PubChem CID 111634
ChEBI CHEBI:33068
IUPAC Name 1H-indol-7-amine
SMILES C1=CC2=C(C(=C1)N)NC=C2

Specifications

Melting Point 90°C to 91°C
Quantity 100 mg
Beilstein 471787
Sensitivity Air sensitive
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 7-Aminoindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.