Learn More
7-Aminoindole, 97%
Product Code. 15479069
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 15479069
Brand: Thermo Scientific Alfa Aesar L17794.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 5192-04-1 | C8H8N2 | 132.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 5192-04-1
Melting Point 90°C to 91°C
Molecular Formula C8H8N2
MDL Number MFCD02093960
Quantity 1 g
Beilstein 471787
Sensitivity Air sensitive
Synonym 7-aminoindole, 7-amino-1 h-indole, 1 h-indol-7-ylamine, 7-indolamine, indole-7-ylamine
InChI Key WTFWZOSMUGZKNZ-UHFFFAOYSA-N
SMILES NC1=C2NC=CC2=CC=C1
IUPAC Name 1H-indol-7-amine
Molecular Weight (g/mol) 132.17
PubChem CID 111634
ChEBI CHEBI:33068
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 7-Aminoindole