Learn More
(7,8,9,10-Tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxalin-2-yl)methanol, TRC
Specifications
Specifications
| Chemical Name or Material | (7,8,9,10-Tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxalin-2-yl)methanol |
| CAS | 2832128-82-0 |
| Molecular Formula | C14 H15 N3 O |
| Quantity | 1 mg |
| Synonym | Hydroxymethyl Varenicline |
| InChI Formula | InChI=1S/C14H15N3O/c18-7-10-6-16-13-2-11-8-1-9(5-15-4-8)12(11)3-14(13)17-10/h2-3,6,8-9,15,18H,1,4-5,7H2 |
| SMILES | OCc1cnc2cc3C4CNCC(C4)c3cc2n1 |
| IUPAC Name | 5,8,14-triazatetracyclo[10.3.1.02,11.04,9]hexadeca-2(11),3,5,7,9-pentaen-6-ylmethanol |
| Molecular Weight (g/mol) | 241.288 |
| Formula Weight | 241.122 |