Learn More
DiscoverX 7α,25-dihydroxycholesterol
Product Code. 17352509
Encompass
Change view
Click to view available options
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17352509 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17352509 Supplier DiscoverX Supplier No. 921186
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
7α,25-dihydroxycholesterol is an agonist for GPR183. Provided as 5mg of dry powder.
7α,25-dihydroxycholesterol (7α,25-DHC) is a potent and selective agonist of the G protein-coupled receptor GPR183 (KD=450pM). Acts as a potent chemokine, directing migration of B,T, and dendritic cells, and plays an important role in adaptive immune response. Provided as 5mg of dry powder.
Specifications
Quantity 5 mg