Learn More
DiscoverX 7α,25-dihydroxycholesterol
Product Code. 17352509
Encompass
Change view
Click to view available options
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17352509 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17352509 Supplier DiscoverX Supplier No. 921186
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
7α,25-dihydroxycholesterol is an agonist for GPR183. Provided as 5mg of dry powder.
7α,25-dihydroxycholesterol (7α,25-DHC) is a potent and selective agonist of the G protein-coupled receptor GPR183 (KD=450pM). Acts as a potent chemokine, directing migration of B,T, and dendritic cells, and plays an important role in adaptive immune response. Provided as 5mg of dry powder.
Specifications
Quantity 5 mg