Learn More
(6Z)-8-Methyl-6-nonenoic Acid (up to 10% trans isomer), TRC
Product Code. 30466881
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466881 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30466881 Supplier Toronto Research Chemicals Supplier No. TRCM326245100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (6Z)-8-Methyl-6-nonenoic Acid
CAS 31467-60-4
Molecular Formula C10 H18 O2
Quantity 100 mg
Synonym (Z)-8-Methyl-6-Nonenoic Acid, 8-Methyl-cis-6-nonenoic Acid, cis-8-Methyl-6-nonenoic Acid
InChI Formula InChI=1S/C10H18O2/c1-9(2)7-5-3-4-6-8-10(11)12/h5,7,9H,3-4,6,8H2,1-2H3,(H,11,12)/b7-5-
SMILES CC(C)\C=C/CCCCC(=O)O
IUPAC Name (Z)-8-methylnon-6-enoic acid
Molecular Weight (g/mol) 170.25
Formula Weight 170.1307