missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1983BHRP
297.60 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00900kg
Description
PA1-983B-HRP detects His-tagged proteins in Western Blot applications.
6x-His Tag Polyclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for Western Blot.Specifications
6x-His Tag | |
Polyclonal | |
HRP | |
6His tag; 6X His; His Tag; Histidine Tag | |
Synthetic peptide corresponding to residues H H H H H H | |
50 μL | |
Primary | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.07% Kathon | |
Rabbit | |
Affinity chromatography | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |