Learn More
6PPD-quinone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-((4-Methylpentan-2-yl)amino)-5-(phenylamino)cyclohexa-2,5-diene-1,4-dione |
| CAS | 2754428-18-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H22 N2 O2 |
| Quantity | 5 mg |
| Synonym | 2-((4-Methylpentan-2-yl)amino)-5-(phenylamino)cyclohexa-2,5-diene-1,4-dione, 6PPD-quinone, 6PPD-Quinone, 2-[(1,3-Dimethylbutyl)amino]-5-(phenylamino)-2,5-cyclohexadiene-1,4-dione (ACI), 2,5-Cyclohexadiene-1,4-dione','2-[(1,3-dimethylbutyl)amino]-5-(phenylamino)- (ACI) |
| InChI Formula | InChI=1S/C18H22N2O2/c1-12(2)9-13(3)19-15-10-18(22)16(11-17(15)21)20-14-7-5-4-6-8-14/h4-8,10-13,19-20H,9H2,1-3H3 |
| SMILES | CC(C)CC(C)NC1=CC(=O)C(=CC1=O)Nc2ccccc2 |
| IUPAC Name | 2-anilino-5-(4-methylpentan-2-ylamino)cyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 298.38 |
| Show More |