Learn More
6PPD-quinone-D5, TRC
Specifications
Specifications
| Chemical Name or Material | 2-((4-Methylpentan-2-yl)amino)-5-(phenylamino)cyclohexa-2,5-diene-1,4-dione D5 (Aniline D5) |
| CAS | 2750119-14-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 D5 H17 N2 O2 |
| Quantity | 5 mg |
| Synonym | 2-((4-Methylpentan-2-yl)amino)-5-(phenylamino)cyclohexa-2,5-diene-1,4-dione D5 (Aniline D5), 6PPD-quinone D5, 6PPD-Quinone D5 (Aniline D5), 2-((4-Methylpentan-2-yl)amino)-5-(phenylamino)cyclohexa-2,5-diene-1,4-dione (6PPD-Quinone) D5 |
| InChI Formula | InChI=1S/C18H22N2O2/c1-12(2)9-13(3)19-15-10-18(22)16(11-17(15)21)20-14-7-5-4-6-8-14/h4-8,10-13,19-20H,9H2,1-3H3/i4D,5D,6D,7D,8D |
| SMILES | [2H]c1c([2H])c([2H])c(NC2=CC(=O)C(=CC2=O)NC(C)CC(C)C)c([2H])c1[2H] |
| IUPAC Name | 2-(4-methylpentan-2-ylamino)-5-(2,3,4,5,6-pentadeuterioanilino)cyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 303.41 |
| Show More |