Learn More
Anton Paar 600ML SILICONE VISCOSITY STANDARD AP RT500 - 600 ml - 554 / 501 mPa.s at 20 / 25 °C
Brand: Anton Paar 191086
600ML SILICONE VISCOSITY STANDARD AP RT500 - 600 ml - 554 / 501 mPa.s at 20 / 25 °C