Learn More
6-tert-Butyldimethylsilyl-4’-hydroxy Raloxifene-d4, TRC
Product Code. 30346305
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346305 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30346305 Supplier Toronto Research Chemicals Supplier No. TRCB6913422.5MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-tert-Butyldimethylsilyl-4'-hydroxy Raloxifene-d4
CAS 1329611-25-7
Molecular Formula C34H37D4NO4SSi
Quantity 2.5 mg
Synonym [6-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-2-(4-hydroxyphenyl)benzo[b]thien-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone-d4
InChI Formula InChI=1S/C34H41NO4SSi/c1-34(2,3)41(4,5)39-28-17-18-29-30(23-28)40-33(25-9-13-26(36)14-10-25)31(29)32(37)24-11-15-27(16-12-24)38-22-21-35-19-7-6-8-20-35/h9-18,23,36H,6-8,19-22H2,1-5H3/i21D2,22D2
SMILES [2H]C([2H])(Oc1ccc(cc1)C(=O)c2c(sc3cc(O[Si](C)(C)C(C)(C)C)ccc23)c4ccc(O)cc4)C([2H])([2H])N5CCCCC5
IUPAC Name [6-[tert-butyl(dimethyl)silyl]oxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(1,1,2,2-tetradeuterio-2-piperidin-1-ylethoxy)phenyl]methanone
Molecular Weight (g/mol) 591.87
Formula Weight 591.2777