Learn More
6-Methylpyridine-3-amine, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Methylpyridine-3-amine |
| CAS | 3430-14-6 |
| Molecular Formula | C6H8N2 |
| Quantity | 1 g |
| Synonym | 5-Amino-2-picoline, 2-Methyl-5-aminopyridine, 3-Amino-6-methylpyridine, 5-Amino-2-methylpyridine, 5-Amino-2-picoline, 6-Methylpyridin-3-ylamine, 6-Methylpyridine-3-amine, NSC 52872 |
| InChI Formula | InChI=1S/C6H8N2/c1-5-2-3-6(7)4-8-5/h2-4H,7H2,1H3 |
| SMILES | Cc1ccc(N)cn1 |
| IUPAC Name | 6-methylpyridin-3-amine |
| Molecular Weight (g/mol) | 108.14 |
| Formula Weight | 108.0687 |