Learn More
6-Methylene Progesterone Acetate, TRC
Product Code. 30137719
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137719 100 mg 100mg
30136008 50 mg 50mg
30010690 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137719 Supplier Toronto Research Chemicals Supplier No. TRCM304220100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32634-95-0
Molecular Formula C24 H32 O4
Molecular Weight (g/mol) 384.51
Synonym 6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate, Medroxyprogesterone Acetate Imp. E (EP), 6-Methylenehydroxyprogesterone Acetate, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methylene-, Pregn-4-ene-3,20-dione, 17-hydroxy-6-methylene-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6-methylene-, acetate (6CI), 17-(Acetyloxy)-6-methylenepregn-4-ene-3,20-dione, 17-Acetoxy-6-methylenepregn-4-ene-3,20-dione, 17α-Acetoxy-6-methylenepregn-4-ene-3,20-dione, 17α-Acetoxy-6-methyleneprogesterone, 6,6'-Dehydromedroxyprogesterone acetate, NSC 58799
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-6-methylidene-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CC(=C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13,18-20H,1,6-12H2,2-5H3/t18-,19+,20+,22-,23+,24+/m1/s1
Formula Weight 384.2301
Chemical Name or Material 6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate (6-Methylenehydroxyprogesterone Acetate)