Learn More
6-Methyl-5-nitroquinoline, 97%, Thermo Scientific™
Product Code. 11370234
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11370234 5 g 5g
11360234 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11370234 Supplier Thermo Scientific Alfa Aesar Supplier No. L00807.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23141-61-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00024021
InChI Key CENBTULRDDPOGR-UHFFFAOYSA-N
Synonym 5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
PubChem CID 90009
IUPAC Name 6-methyl-5-nitroquinoline
SMILES CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]
Specifications
Melting Point 115°C to 118°C
Quantity 5 g
Beilstein 154688
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 6-Methyl-5-nitroquinoline