Learn More
6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carboxylic Acid |
| CAS | 10176-79-1 |
| Molecular Formula | C13H11NO3 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C13H11NO3/c1-8-10(9-5-3-2-4-6-9)7-11(13(16)17)12(15)14-8/h2-7H,1H3,(H,14,15)(H,16,17) |
| SMILES | CC1=C(C=C(C(=O)O)C(=O)N1)c2ccccc2 |
| IUPAC Name | 6-methyl-2-oxo-5-phenyl-1H-pyridine-3-carboxylic acid |
| Molecular Weight (g/mol) | 229.23 |
| Formula Weight | 229.0739 |