Learn More
6-Fluoroisatin, TRC
Product Code. 30391971
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391971 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30391971 Supplier Toronto Research Chemicals Supplier No. TRCF5920385G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Fluoroisatin
CAS 324-03-8
Molecular Formula C8H4FNO2
Quantity 5 g
InChI Formula InChI=1S/C8H4FNO2/c9-4-1-2-5-6(3-4)10-8(12)7(5)11/h1-3H,(H,10,11,12)
SMILES FC1=CC(NC(C2=O)=O)=C2C=C1
IUPAC Name 6-fluoroindoline-2,3-dione
Molecular Weight (g/mol) 165.12
Formula Weight 165.0226