missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro L-DOPA Hydrobromide Salt, TRC

Product Code. 30390744
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390744 50 mg
30406332 10 mg
30404771 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390744 Supplier Toronto Research Chemicals Supplier No. TRCF59154650MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154051-94-2
Molecular Formula C9 H10 F N O4 . Br H
Molecular Weight (g/mol) 296.09
Synonym L-Tyrosine, 2-fluoro-5-hydroxy-, hydrobromide (9CI), 6-Fluoro-L-dopa hydrobromide
IUPAC Name (2S)-2-amino-3-(2-fluoro-4,5-dihydroxyphenyl)propanoic acid;hydrobromide
SMILES Br.N[C@@H](Cc1cc(O)c(O)cc1F)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H10FNO4.BrH/c10-5-3-8(13)7(12)2-4(5)1-6(11)9(14)15;/h2-3,6,12-13H,1,11H2,(H,14,15);1H/t6-;/m0./s1
Formula Weight 294.9855
Percent Purity >95%
Chemical Name or Material 6-Fluoro-L-dopa hydrobromide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.