Learn More
6-Fluoro-8-Quinolinesulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Fluoro-8-Quinolinesulfonyl Chloride |
| CAS | 1016515-89-1 |
| Molecular Formula | C9H5ClFNO2S |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C9H5ClFNO2S/c10-15(13,14)8-5-7(11)4-6-2-1-3-12-9(6)8/h1-5H |
| SMILES | O=S(=O)(Cl)C1=CC(F)=CC2=CC=CN=C21 |
| IUPAC Name | 6-fluoroquinoline-8-sulfonyl chloride |
| Molecular Weight (g/mol) | 245.66 |
| Formula Weight | 244.97 |