Learn More
6-Ethylchrysene, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Ethylchrysene |
| CAS | 2732-58-3 |
| Molecular Formula | C20 H16 |
| Quantity | 50 mg |
| Synonym | 6-Ethyl-chrysene |
| InChI Formula | InChI=1S/C20H16/c1-2-14-13-20-17-9-4-3-7-15(17)11-12-19(20)18-10-6-5-8-16(14)18/h3-13H,2H2,1H3 |
| SMILES | CCc1cc2c3ccccc3ccc2c4ccccc14 |
| IUPAC Name | 6-ethylchrysene |
| Molecular Weight (g/mol) | 256.341 |
| Formula Weight | 256.125 |