Learn More
6-Des(1-methyl-2-benzimidazolyl)-6-carboxy Telmisartan, TRC
Product Code. 30349396
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349396 10 mg 10mg
30358911 50 mg 50mg
30359124 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30349396 Supplier Toronto Research Chemicals Supplier No. TRCD29102010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 884330-12-5
Molecular Formula C26 H24 N2 O4
Molecular Weight (g/mol) 428.4798
Synonym 1-[(2'-Carboxy[1,1'-biphenyl]-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazole-6-carboxylic acid, 1-[(2'-Carboxybiphenyl-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazol-6-carboxylic Acid, 1H-Benzimidazole-6-carboxylic acid, 1-[(2'-carboxy[1,1'-biphenyl]-4-yl)methyl]-4-methyl-2-propyl-, Telmisartan Imp E (EP)
IUPAC Name 3-[[4-(2-carboxyphenyl)phenyl]methyl]-7-methyl-2-propylbenzimidazole-5-carboxylic acid
SMILES CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)O)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C26H24N2O4/c1-3-6-23-27-24-16(2)13-19(25(29)30)14-22(24)28(23)15-17-9-11-18(12-10-17)20-7-4-5-8-21(20)26(31)32/h4-5,7-14H,3,6,15H2,1-2H3,(H,29,30)(H,31,32)
Formula Weight 428.1736
Percent Purity >95%
Chemical Name or Material 1-[(2'-Carboxybiphenyl-4-yl)methyl]-4-methyl-2-propyl-1H-benzimidazol-6-carboxylic Acid
Grade HPLC