Learn More
[(η-6-Cymene)[(S,S)-1,2-diphenyl-N-tosyl-1,2-ethanediaminato]ruthenium] chloride], TRC
Product Code. 30365876
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30365876 Supplier Toronto Research Chemicals Supplier No. TRCC989475100MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material [(eta-6-Cymene)[(S,S)-1,2-diphenyl-N-tosyl-1,2-ethanediaminato]ruthenium] chloride]
CAS 192139-90-5
Molecular Formula C31H35ClN2O2RuS
Quantity 100 mg
Synonym [RuCl(TsDPEN)(η-6-cymene)], [((S,S)-2-Amino-1,2-diphenylethyl)[(4-tolyl)sulfonyl]amido](chloro)(η6-p-cymene)ruthenium, , [S-(R*,R*)]-[N-[2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium, [N-[(1S,2S)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium
InChI Formula InChI=1S/C21H21N2O2S.C10H14.ClH.Ru/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17;1-8(2)10-6-4-9(3)5-7-10;;/h2-15,20-21H,22H2,1H3;4-8H,1-3H3;1H;/q-1;;;+2/p-1/t20-,21-;;;/m0.../s1
SMILES [Cl-][Ru+2]123456(C=7(C(C)C)[CH]1=[CH]2C3(C)=[CH]4[CH]57)[N-](S(=O)(=O)C8=CC=C(C)C=C8)[C@H]([C@@H]([NH2]6)C9=CC=CC=C9)C%10=CC=CC=C%10
Molecular Weight (g/mol) 636.21
Formula Weight 636.12